Documenti fondamentali
22027
Swagelok® Front Ferrule
Swagelok® 403-1, stainless steel, 1/4 in. Swagelok
Sinonimo/i:
Swagelok® 403-1,SS FRT FERR 1/4" PK5
About This Item
Codice UNSPSC:
41115717
Materiali
stainless steel
Confezionamento
pkg of 5 ea
Produttore/marchio commerciale
Swagelok® 403-1
Raccordi
1/4 in. Swagelok
Descrizione generale
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Brass ferrules are used with copper tubing whereas stainless-steel ferrules are used with stainless-steel tubing. Furthermore, the stainless steel ferrules have great mechanical strength but they cannot be removed once they are on the column.
Note legali
Swagelok is a registered trademark of Swagelok Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Gas Chromatographic Reactors.
Patton JE.
Gas Chromatography, 1, 133-133 (1971)
Grob RL & Barry EF.
Modern Practice of Gas Chromatography, 522-523 (2004)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)