Z756946
Universal animal restrainer
large, pack of 1 ea
Sinonimo/i:
animal container
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.22
Prodotti consigliati
Materiali
clear acrylic
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art H46400-0003
Descrizione generale
Holds animal: guinea pigs
A variety of small animals and rodents can be restrained with this unit simply by moving the head gate to one of 4 different positions. To accommodate larger rodents, simply pull out the inner floor, increasing the available room. Stable base legs are provided by milled, chrome-plated brass thumb screws
Minimum restraining: 15.2 x 7.3 cm (6 x 2 7/8 in.)
Maximum restraining: 21.6 x 7.3 cm (8 1/2 x 2 7/8 in.)
A variety of small animals and rodents can be restrained with this unit simply by moving the head gate to one of 4 different positions. To accommodate larger rodents, simply pull out the inner floor, increasing the available room. Stable base legs are provided by milled, chrome-plated brass thumb screws
- Humanely restrain small animals
- Comes with vented head gate and tail gate that are easily removed
- Occlusion cuff held close to the animal′s body with the tailgate
Minimum restraining: 15.2 x 7.3 cm (6 x 2 7/8 in.)
Maximum restraining: 21.6 x 7.3 cm (8 1/2 x 2 7/8 in.)
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)