Documenti fondamentali
Z756709
Scienceware® 50 mL tube holder
grip style, round blue polystyrene
Sinonimo/i:
centrifuge tube holder, tube rack
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.51
Materiali
round blue polystyrene
Caratteristiche
Holds 1 x 50 mL tube
Confezionamento
bag of 5 ea
Produttore/marchio commerciale
Bel-Art F18795-0001
Descrizione generale
Leave both hands free for pipetting and other critical tasks. The Grip Style gently grips the tube and moves with the tube when it is lifted from the bench top.
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)