Documenti fondamentali
Z744128
Tough-Tags dispensing station
holds 5 boxes of Tough-Tags
Sinonimo/i:
solvent resistant labels, temperature resistant labels
About This Item
Codice UNSPSC:
55121611
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Diversified Biotech TTSA-9000
Descrizione generale
Tough-Tag Station efficiently dispenses labels.
  • Top-Loading - Easily insert new rolls or remove empty boxes
  • Secure - Specially designed top rail holds rolls in place
  • Cutting Edge - Allows easy liner trimming
  • Convenient - When loaded, station can be used as a writing surface
  • Versatile - Also holds 5-Across Tough-Spots®,Bake-Tags, Teeny Tough-Tags and Sidewall Tough-Tags!
Caratteristiche e vantaggi
Versatile holds 5-Across Tough-Spots®, Bake-Tags, Teeny Tough-Tags and Sidewall Tough-Tags
Note legali
Tough-Spots is a registered trademark of Diversified Biotech
Tough-Tags is a trademark of Diversified Biotech
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)