Z740418
Specimen containers with snap cap
size 500 mL
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Prodotti consigliati
Materiali
polypropylene cap
polypropylene container
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Simport C580-4
Misura
500 mL
Descrizione generale
These disposable containers made of polypropylene, are ideal for collection and storage of fluids, powders, solids, pathology specimens and hazardous samples. Lids are tight fitting
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)