Documenti fondamentali
Z738603
Glow Writer autoradiography pen
phosphorescent marking pen
About This Item
Codice UNSPSC:
41105341
NACRES:
NB.22
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Diversified Biotech GLWR-1000
Descrizione generale
Glow Writer is a phosphorescent marking pen for annotating or marking chemiluminescent and isotope-labeled substrates for X-ray film autography. The unique marker delivers a phosphor-containing marking ink through a fine point porous nib. Before exposure to X-ray film, the phosphorescent writing is excited by bright room light. A phosphor ink having an optimum concentration of phosphor and a relatively short afterglow half-life (approx. 2-3 minutes) has been selected so that upon exposure to X-ray film, autographic exposures of almost any duration will be easily readable on the film. The fine point nib is easily replaceable if it dries out.
Note legali
Glow Writer is a trademark of Diversified Biotech
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)