Z719935
Spinbar® Capsule Magnetic Stirring Bar
pkg of 1 ea
Sinonimo/i:
Stir Bar, Stirbar, Stirring Bar
About This Item
Codice UNSPSC:
41103806
NACRES:
NB.72
Prodotti consigliati
Materiali
polygon PTFE-coated (in flat housing)
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art F37152-0000
Ancoretta: lungh. × diam.
1.3 cm × 5.3 cm
Descrizione generale
Designed for stirring in awkward vessels, for gently mixing staining solutions above delicate gels, or for quickly dissolving chemicals. A small flat capsule contains the magnetic stirring bar that spins freely inside without contact with the vessel bottom or gel. The protective capsule is molded with polystyrene and shelters a Spinbar magnetic stirring bar which is molded with inert PTFE. Bar diameter is 1 3/16 in.
Note legali
Spinbar is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)