Z709549
Sartorius mLINE® mechanical pipettor
Single channel, volume 10-100 μL
Sinonimo/i:
Biohit pipettes, pipettor
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prodotti consigliati
Caratteristiche
electronic: no
single-channel
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Sartorius 725050
Campo di misura del volume
10-100 μL
Volume
100 μL
Descrizione generale
Sartorius′s new mechanical pipettor family - the mLINE - offers a unique combination of novel features for mechanical pipetting. Excellent performance, full autoclavability, smooth ergonomics and perfect safety. The mLINE pipettors cover the full volume range of 0.1 to 5000.
- Extremely light pipetting and tip ejection
- Unique safety features
- Stepless volume adjustment and patented volume locking system
- Thermal insulation of internal components to improve accuracy
- Full autoclavability without disassembling
- Easy cleaning, maintenance and calibration
- ID tag and color-coded caps
- Safe-Cone Filters and unique ejection
- Opti-load tip mounting mechanism for even tip sealing and light tip ejection in multichannel pipettors
Note legali
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)