Documenti fondamentali
Z694533
Nanofiber multiwell plate, with aligned nanofibers
size 24 wells
Sinonimo/i:
24 well culture plates, 3D, 3D Cell Culture, 3d plates, tissue culture plates
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.13
Materiali
(polycaprolactone (PCL))
flat clear wells
transparent polycaprolactone (PCL)
Sterilità
sterile
Caratteristiche
lid
Produttore/marchio commerciale
Nanofiber Solutions 2402
Misura
24 wells
Compatibilità
suitable for (Tissue engineering, cancer research)
Tipo di binding
non-treated surface
Descrizione generale
Nanofiber cell culture dishes, multiwell plates and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for cancer research. The cells can be simply removed for protein or gene analysis.
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
For fluorescent imaging applications, we recommend using the Nanofiber Solutions chamber slides.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Articoli
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models