Documenti fondamentali
Z681784
Culture tubes
size 25 mm × 150 mm , closure caps are sold separately, borosilicate glass tube
Sinonimo/i:
culture tube, test tube, test tubes
About This Item
Codice UNSPSC:
41121706
NACRES:
NB.22
Materiali
borosilicate glass tube
conical cap (Push fit)
round bottom
straight neck
transparent tube
Sterilità
non-sterile
Caratteristiche
neck thread push fit
Confezionamento
case of 288 ea (4 x 72)
pack of 72 ea
Produttore/marchio commerciale
Kimble® 45048-25150
Capacità
55 mL
Misura
25 mm × 150 mm , closure caps are sold separately
Descrizione generale
KIMAX® culture tube has a fire-polished top rim and uniform wall thickness for maximum heat transfer and chemical resistance.
Caratteristiche e vantaggi
  • Kimble® culture tubes are manufactured from low extractable, chemical-resistant borosilicate glass.
  • These reusable tubes feature a fire-polished top rim.
  • Uniform wall thickness ensures maximum heat transfer.
  • The marking spot is printed in durable white ceramic enamel.
Confezionamento
Packaged in a sturdy cardboard box with internal grid to minimize damage during shipping and handling.
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)