Z673420
KDS stainless steel syringes
volume 50 mL, pressure 700-1500 psi
Sinonimo/i:
stainless steel syringes
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prodotti consigliati
Descrizione
with Luer lock
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
KD Scientific 780817
Parametri
700-1500 psi pressure
Lungh. × D.E.
5.49 in. (139.3 mm) × 1.25 in. (31.8 mm)
Volume
50 mL
Descrizione generale
These rugged #316 stainless steel syringes are a perfect complement to the KDS 410 high pressure syringe dispenser. They eliminate the problem of breaking glass syringes when using high pressures. Compatibility with most chemicals is assured as wetted parts are #316 stainless steel, Viton® for the 20, 50 and 100 mL sizes, and Perfluoroelastomer for the 8 mL size.
Caratteristiche e vantaggi
- Compatible with most syringe pumps
- Fully autoclavable
- Good resistance to most chemicals
- Safe solution when using high pressure dispensing
Note legali
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)