Documenti fondamentali
Z654310
Caps for Duran® laboratory bottles
GL 45 neck thread, colorless premium cap TPCh260 (with PTFE coated seal)
Sinonimo/i:
caps bottles, glass bottles caps
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
colorless premium cap TPCh260 (with PTFE coated seal)
Produttore/marchio commerciale
DURAN®
Schott
Parametri
-196-260 °C temp. stability
Raccordi
GL 45 neck thread
Descrizione generale
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Note legali
DURAN is a registered trademark of DWK Life Sciences