Documenti fondamentali
Z614068
Vivaspin 500 centrifugal concentrators
MWCO 30 kDa
Sinonimo/i:
Vivaspin 500
About This Item
Codice UNSPSC:
41104916
NACRES:
NB.24
Materiali
polycarbonate housing
polyethersulfone membrane
Descrizione
PES membrane
Sterilità
non-sterile
Caratteristiche
holdup volume 5 μL
Confezionamento
package of 100 ea
Produttore/marchio commerciale
Sartorius VS0122
Parametri
15000 × g max. centrifugal force
Tasca dead stop
5 μL
Area membrana
0.5 cm2
Dimens. tubo
~2.2 mL
Volume d’esercizio
500 μL
Dimensione pori
30 kDa MWCO
Descrizione generale
  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)