Z370789
Scienceware® cloning cylinders, polystyrene
40 of each size
Sinonimo/i:
cloning cylinder, cloning equipment
About This Item
Codice UNSPSC:
41106618
NACRES:
NB.22
Prodotti consigliati
Sterilità
sterile
Confezionamento
box of 3 bags
Produttore/marchio commerciale
Bel-Art H37847-0000
Descrizione generale
To isolate a clone, sterile grease (not included) is applied to bottom edge of cylinder and inverted over clone of choice. Provides an isolation chamber to trypsinize and recover clonal colonies.
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)