Z370770
Scienceware® Holdfast™ graduated cylinders with handle
volume 2000 mL
About This Item
Codice UNSPSC:
41121805
NACRES:
NB.43
Prodotti consigliati
Materiali
polypropylene
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F28461-2000
Campo di misura del volume
2000 mL
Descrizione generale
Autoclavable polypropylene with double scale and stable octagon base. The handle provides a safe, solid grip for more controlled pouring from these large-sized cylinders.
Note legali
Holdfast is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)