WPB01402WA
Whirl-Pak® Stand-Up Thio-Bag®, Waste Water Sampling Bag
100 mL capacity
About This Item
Codice UNSPSC:
24111500
NACRES:
NB.35
Prodotti consigliati
Sterilità
sterile
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
Nasco B01402WA
Parametri
-346-180 °F temp. range (-210 to 82 °C)
Largh. × lungh. × spess.
7.5 cm × 18.5 cm × 0.064 mm
Capacità
100 mL
100 mL capacity
Categorie correlate
Descrizione generale
A special gusset on the bottom of the bag flattens out when water is put in the bag, allowing it to stand up completely by itself. No rack or other holder is needed to keep the bag upright. Forget washing and sterilizing glass bottles and disposing of expensive vials. Sterile and unbreakable, these bags will save time and money whether working in the lab or in the field. Each bag contains active sodium thiosulfate to neutralize chlorine in the sample.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen, temperatures down to -346°F (-210°C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen, temperatures down to -346°F (-210°C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Note legali
Thio-Bag is a registered trademark of The Aristotle Corporation
Whirl-pak is a registered trademark of The Aristotle Corporation
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
WPB01402WA-100EA | 4061833576519 |