Documenti fondamentali
WHAWB100082
Unicore Punch Kit
I.D. 6.00 mm, pkg of 25 ea
Sinonimo/i:
UniCore Punches 6.00 mm (25), Whatman FTA, Z708909, harris punch, harris uni-core, uni-core, whatman punch, Harris Uni-Core
About This Item
Codice UNSPSC:
41105300
NACRES:
NB.22
Confezionamento
pkg of 25 ea
Produttore/marchio commerciale
Qiagen WB100082
Diam. int.
6.00 mm
Descrizione generale
An inexpensive disposable multi-purpose coring tool designed for simultaneous sample extraction, collection and storage in one operation. Suitably samples thin and soft substrates including filter paper, gels and dried blood on cards. Units can be autoclaved at 250°C for limited re-use.
Manual UniCore punch system for sample disc removal from FTA cards.
Caratteristiche e vantaggi
  • Stainless steel cutting tip (ranging from 6.0 mm) enables manual coring, retrieval, and controlled ejection of the FTA punch.
  • Designed to minimize contamination and carryover between samples when a cleaning procedure is followed.
Altre note
Field of Use : For internal research use only
Note legali
FTA is a trademark of Qiagen Group
Qiagen is a registered trademark of Qiagen Group
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Unicore Punch Kit I.D. 1.2 mm, pkg of 25 ea
WHAWB100074
Unicore Punch Kit
Protocolli
This document shows intructions for EasiCollect device from GE Healthcare