WHA77002804
Whatman® UNIFILTER® 96 well filter plates
25-30 μm, clear polystyrene, melt brown polypropylene membrane, non-sterile, biomolecule, 25 ea
Sinonimo/i:
Z382345, micro plate, multiwell plate, well plates
About This Item
Prodotti consigliati
Nome del prodotto
Whatman® UNIFILTER® plates, working volume 800 μL, with 25-30μm MBPP membrane
Materiali
clear polystyrene
conical bottom clear wells
polystyrene filter
Sterilità
non-sterile
Confezionamento
pkg of 25 ea
Produttore/marchio commerciale
Cytiva
Whatman 7700-2804
Whatman Article No. 28420807 (US reference)
Volume massimo
800 μL
Misura
96 wells
Volume d’esercizio
800 μL
Dimensione pori
25-30 μm
Tipo di binding
non-treated surface
Categorie correlate
Caratteristiche e vantaggi
- No cross talk. Patented integral filter design prevents well-to-well cross contamination.
- Economical to use. Wide range of well volume options ensures efficient use of materials.
- Better control. Choice of filter media allows control of the flow rates and retention characteristics.
- Versatile. A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose.
Altre note
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
WHA77002804 | 4061826077405 |