TMO4116-2000
Nalgene® sterile, single-use Erlenmeyer flask with vented closure
PETG, bottle volume (2000 mL), baffled bottom, case of 4 ea
Sinonimo/i:
Nalgene Erlenmeyer flask, single use plastic flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.13
Prodotti consigliati
Materiali
PETG
Sterilità
sterile
Caratteristiche
vented
Confezionamento
case of 4 ea
Produttore/marchio commerciale
Nalgene 4116-2000
Volume del flacone
(2000 mL)
Descrizione generale
Save preparation time and avoid contamination risk by using the Nalgene® Single-Use PETG Erlenmeyer Flasks, which are available with a flat or baffled bottom. These flasks are ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available with a vented or non-vented closure.
Caratteristiche e vantaggi
- Sterile and single us, reducing contamination risk
- BPA-free
- Collapses when autoclaved, reducing bio hazardous waste
- Graduated for quick volume assessment
- Increases user safety due to the break-resistant and robust design of the crystal-clear PETG material
- Facilitates visual inspection and monitoring of contents due to its crystal-clear composition
- Equipped with a vented cap to support gas exchange during processes like cell culture
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)