TMO4113-2000
Nalgene® sterile single-use Erlenmeyer flask with solid closure
PETG, bottle volume 2000 mL, baffled bottom, case of 4 ea
Sinonimo/i:
Nalgene Erlenmeyer flask, single use plastic flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.13
Materiali
PETG
Sterilità
sterile
Confezionamento
case of 4 ea
Produttore/marchio commerciale
Nalgene 4113-2000
Volume del flacone
2000 mL
Descrizione generale
Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Altre note
Browse all Erlenmeyer flasks
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)