Documenti fondamentali
TMO362150-1280
Nalgene® white caps
polypropylene, diam. × thread 30 mm × 28-415 mm, case of 500 ea
Sinonimo/i:
leakproof closure, threaded cap
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
polypropylene
white
Confezionamento
case of 500 ea
Produttore/marchio commerciale
Nalgene 362150-1280
Diam. × filettatura
30 mm × 28-415 mm
Descrizione generale
Eliminate spills with Thermo Scientific Nalgene Colored Polypropylene Closures. These one-piece linerless closures fit Nalgene narrow-mouth 4mL bottles. They feature an integrally molded seal ring and a well-defined closure knurl that makes gripping and capping easy.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)