TMO362002-0004
Nalgene® Boston round bottle without cap
natural high-density polyethylene, volume 125 mL, case of 500 ea
Sinonimo/i:
HDPE bottle, nalgene bottle
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
natural high-density polyethylene
Confezionamento
case of 500 ea
Produttore/marchio commerciale
Nalgene 362002-0004
Capacità
125 mL
Volume
125 mL
Categorie correlate
Descrizione generale
Conduct demanding packaging applications with Thermo Scientific Nalgene Boston Round HDPE Bottles without Closure, which are both durable and reliable products. These high density polyethylene bottles are rigid and translucent with excellent chemical resistance. Uniform walls make these bottles resistant to splitting and puncturing.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)