TMO312018-0500
Nalgene® square graduated bottle
high-density polyethylene, bottle capacity 500 mL, natural, case of 250 ea
Sinonimo/i:
break resistant bottle, chemcial resistant bottle
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
high-density polyethylene
natural
square
Confezionamento
case of 250 ea
Produttore/marchio commerciale
Nalgene 312018-0500
Capacità bottiglia
500 mL
Categorie correlate
Descrizione generale
Save space with chemical- and break-resistant Thermo Scientific Nalgene Square HDPE Graduated Bottles with Closure. These translucent bottles provide visible liquid levels and molded-in graduations for easy measuring. Square bottles with polypropylene closures feature space-saving designs and drip-proof necks for convenient dispensing.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)