TMO241205
Nunc® breathable cell culture sealing film
rayon acrylate, sterile, case of 50 ea
Sinonimo/i:
adhesive film, adhesive foil
About This Item
Codice UNSPSC:
41161502
NACRES:
NB.15
Prodotti consigliati
Sterilità
sterile
Confezionamento
case of 50 ea
Produttore/marchio commerciale
Nunc 241205
Categorie correlate
Descrizione generale
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific Nunc Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
TMO241205 | 4061841581246 |