TMO236370
Nunc® sealing tape
polyester, non-sterile, case of 100 ea
Sinonimo/i:
adhesive film, adhesive tape
About This Item
Codice UNSPSC:
41161502
NACRES:
NB.15
Prodotti consigliati
Sterilità
non-sterile
Confezionamento
case of 100 ea
Produttore/marchio commerciale
Nunc 236370
Descrizione generale
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific Nunc Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)