Documenti fondamentali
TMO2116-0060
Nalgene® straight-sided wide-mouth jar
polycarbonate, jar capacity 60 mL, clear, case of 48 ea
Sinonimo/i:
clear plastic container, screw top container
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
clear
polycarbonate
straight
wide-mouth
Confezionamento
case of 48 ea
Produttore/marchio commerciale
Nalgene 2116-0060
Capacità del barattolo
60 mL
Descrizione generale
Collect and store pastes, solids, specimens and lab supplies with window-clear Thermo Scientific Nalgene Straight-side Wide-Mouth Jars. A wide temperature range (-135°C to +135°C) makes these jars an excellent choice for cold room and refrigerated use all the way to the autoclave. These high impact-resistant jars are extremely versatile. Not liquid leakproof.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)