T0443
Pharmed L/S tubing
size 13
Sinonimo/i:
Saint Gobain AY242606, PeristalticPump Tubing
About This Item
Codice UNSPSC:
41105108
NACRES:
NB.35
Prodotti consigliati
Confezionamento
pack of 25 feet
Produttore/marchio commerciale
Tygon
Diam. int.
0.03 in. (0.8 mm)
Descrizione generale
Tubing for MasterFlex™ Peristaltic Pumps, PharMed tubing has been specifically formulated for the MasterFlex™ pump system. Great for tissue and cell work- nontoxic and nonhemolytic. Long service life minimizes risk of fluid exposure; reduces tubing cost and pump downtime.
- Opaque to UV and visible light to protect light-sensitive fluids
- Heat sealable, bondable, and formable
- Extremely low gas permeability
Note legali
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)