R8634
Scienceware® centrifuge tube rack, epoxy-coated steel
20-position, for 15 mL plastic centrifuge tubes
Sinonimo/i:
wire tube holder
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.35
Prodotti consigliati
Caratteristiche
Holds 20 x 18-20 mm tibes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F18788-2003
Misura
12.5 cm × 11 cm × 6 cm
Descrizione generale
- Scienceware® centrifuge tube racks are made of epoxy-coated steel wire.
- Polygrid racks are resistant to most of the chemicals including organic solvents, salts, weak acids, and weak alkalis.
- Temperature range -223ºC (-369ºF) to 121ºC (250ºF); autoclavable.
Epoxy coated steel wire is chemical and corrosion resistant and steam autoclavable at 121ºC (250ºF).
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)