Documenti fondamentali
R1400
Texan reagent reservoir for multichannel pipettes
with lid, sterile, case of 100 ea (10 packs of 10ea)
Sinonimo/i:
multichannel pipette reservoir, reagent reservoir
About This Item
Codice UNSPSC:
41121522
NACRES:
NB.01
Materiali
V-bottom bottom
colorless
polypropylene
Sterilità
sterile
Confezionamento
case of 100 ea (10 packs of 10ea)
Produttore/marchio commerciale
Excel Scientific RSV-L175-S05
Capacità
175 mL
Descrizione generale
Texan reagent reservoirs are reusable -- an environmentally sound alternative to disposable reagent reservoirs. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Lids are removable -- attached with ball and socket hinges -- and latch tightly for secure storage of left-over reagents. With lids closed, Texan reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 mL.
Note legali
Texan is a trademark of Excel Scientific, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)