Documenti fondamentali
OH30253076
Ohaus® Scout® Portable Balance
Model SKX421, weighing capacity 420 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
About This Item
Codice UNSPSC:
41111502
NACRES:
NB.71
Materiali
metal weighing plate
Caratteristiche
CE compliant
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ohaus® 30253076
Capacità di pesata
420 g, resolution: 0.1 g
Diam. piattino
12 cm
Descrizione generale
Ideal for laboratory and industrial applications, the OHAUS Scout comes in a slim, stackable design with large backlit LCD. Features include superior overload protection, multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high resolution weighing results, these portable balances set a new standard in laboratory and industrial weighing.
Applicazioni
Weighing, Parts Counting, Percent Weighing, Check Weighing, Totalization, Display Hold
Caratteristiche e vantaggi
Stability indicator, stackable design with optional storage cover (excluding 1mg models), mechanical and software overload/underload protection, low battery indicator, auto shut-off, auto tare, span calibration mass included on models up to 620g capacity
Note legali
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)