Key Documents
OH30080693
Ohaus® Conductivity Electrode
STCON7
About This Item
Codice UNSPSC:
39121436
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ohaus® 30080693
Campo di misura della conducibilità
0.02-200 μS/cm
Campo di misura della temperatura
0-100 °C
Descrizione generale
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Applicazioni
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Caratteristiche e vantaggi
Can be used in conjunction with all Starter bench and portable meters
Note legali
Ohaus is a registered trademark of Ohaus Corp.