Documenti fondamentali
OH30031655
Ohaus® Dissolved Oxygen Meter
Model ST300D
About This Item
Codice UNSPSC:
41115611
Materiali
epoxy shaft
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ohaus® 30031655
dissolved oxygen measuring range
0.0-199.9%
Campo di misura della temperatura
0-50 °C
Batteria
4 AAA, 250 hr life
Descrizione generale
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results.
Applicazioni
Dissolved oxygen (DO) measurement
Field and lab Dissolved Oxygen measurement
Caratteristiche e vantaggi
30 measurement memory
99 measurement memory, last calibration data recall, auto barometric pressure measurement, auto temperature compensation with manual salinity compensation
Note legali
Ohaus is a registered trademark of Ohaus Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)