MTCV5809A
MTC™ Bio Cap Inserts for Cryogenic Vials
assorted colors
Sinonimo/i:
cryo storage identification, cryo vial marker, tube storage
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.11
Prodotti consigliati
Materiali
assorted colors
polypropylene
Confezionamento
pack of 500 ea
Produttore/marchio commerciale
MTC™ Bio V5809-A
Descrizione generale
Cap inserts for cryogenic vials, assorted colors, 500/pk
Easily color-code your cryogenic vials for quick visual identification in the lab, for use at temperatures as low as -196°C.
Caratteristiche e vantaggi
- For temperatures as low as -196°C
Note legali
MTC is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)