Documenti fondamentali
MTCP70251S
MTC Bio ASPIR-8 Resevoir for 8-Channel Pipettes
capacity 25 mL, sterile, pack of 100 ea (individually wrapped)
Sinonimo/i:
cryo vial marker, reagent dish, reagent holder
About This Item
Codice UNSPSC:
41121522
NACRES:
NB.15
Sterilità
sterile
Confezionamento
pack of 100 ea (individually wrapped)
Produttore/marchio commerciale
MTC Bio P7025-1S
Capacità
25 mL
Descrizione generale
Aspir-8 Reservoir, 25ml, 8-channel only, sterile, Individually wrapped, 100/cs
Caratteristiche e vantaggi
  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners
Note legali
ASPIR-8 is a trademark of MTC Bio
MTC is a trademark of MTC Bio