Documenti fondamentali
MTCP412311
MTC Bio Transfer Pipette
size 7 mL, large bulb, extended tip, pack of 500 ea (individually wrapped)
Sinonimo/i:
pipet
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.04
Materiali
low-density polyethylene
Caratteristiche
graduation: no
Confezionamento
pack of 500 ea (individually wrapped)
Produttore/marchio commerciale
MTC Bio P4123-11
Lunghezza
150 mm
Capacità bulbo
6.0 mL
Misura
7 mL
Descrizione generale
These unbreakable, one-piece pipettes eliminate contaminated rubber bulbs, broken glass, and exposure to infectious materials. They are made of ultra-clear, low-density polyethylene (LDPE) and are ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Sterile pipettes are excellent for tissue culture applications. Their low-affinity surface reduces sample, cell or protein loss.
Transfer pipette, 7ml, large bulb, extended tip, sterile, individually wrapped, 500/cs
Caratteristiche e vantaggi
  • Transparent, non-toxic and inert
  • Precise, uniform drop size
  • Shatter-proof and unbreakable
  • Heat Sealable
Note legali
MTC is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)