MTCP1001Q
MTC™ Bio Pure.Amp™ qPCR Sealing Film
to fit standard qPCR plates, sticky adhesive
Sinonimo/i:
plastic tube, qPCR sealing membrane, sealing membrane
About This Item
Codice UNSPSC:
41122410
NACRES:
NB.15
Prodotti consigliati
Confezionamento
bag of 10 × 100 ea
Produttore/marchio commerciale
MTC™ Bio P1001-Q
Descrizione generale
Sealing film, qPCR Optical (sticky adhesive film) Bio-Rad Type, 10 bags of 10 seals, 100/pk
Caratteristiche e vantaggi
- RNase, DNase, DNA, and PCR inhibitor free
- Evaporation rates ≤ 5%
- Inner packs of 10 membranes
- -40° to 120°C
Note legali
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)