Documenti fondamentali
HS24670
Lab Alert® four channel timer and clock
Sinonimo/i:
laboratory timer, multichanne timer
About This Item
Codice UNSPSC:
41122411
NACRES:
NB.51
Produttore/marchio commerciale
Heathrow Scientific HS24670
Descrizione generale
Four Channel Timer/Clock
  • Independently or simultaneously set time on four channels
  • Audible and visual alarms signify elapsed time
  • Easily toggle between channels and clock with top buttons
  • Includes spring clip with magnetic back
  • 73 x 59 x 13 mm
Note legali
Lab Alert is a registered trademark of Heathrow Scientific
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)