HS20632C
Pipette Filler
blue natural rubber
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prodotti consigliati
Materiali
blue natural rubber
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Heathrow Scientific HS20632C
Lungh. × Largh. × Alt.
13 cm × 5.4 cm × 5.4 cm
Capacità
~35 mL
Descrizione generale
The natural rubber Pipette Filler is great for use in one-handed operation.
Caratteristiche e vantaggi
- Efficient one-handed operation
- Control air-evacuation, liquid uptake, and liquid dispensing with three valves each containing chemical-resistant stainless steel balls in valves
- Pinch release-valve for smooth liquid flow
- 5.1 in L x 2.1 in W x 2.1 in H (13 cm L x 5.4 cm W x 5.4 cm H)
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)