Documenti fondamentali
HS15996C
Premium slide box
50 place, cork lining, white plastic
Sinonimo/i:
Schiebekasten, slide box microscope, slide storage, slide storage box
About This Item
Codice UNSPSC:
41123302
NACRES:
NB.17
Materiali
nickel closure
white plastic
Caratteristiche
Holds 50 x slides 76x26 mm
Produttore/marchio commerciale
Heathrow Scientific HS15996C
Lungh. × Largh. × Alt.
209 × 86 × 35 mm
Caratteristiche e vantaggi
Slide boxes are perfect for storage or transportation of standard size slides (100 (50 back to back) x 75 mm and 1 x 3").
  • Molded, rigid boxes securely hold slides in grooved slots
  • Durable ABS plastic boxes are available with cork lining only
  • Both the lining and the slots are numbered to correspond to the slide inventory sheet
  • Feature thumb-latch lock mechanism
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)