GE29053612
Tricorn Filter Kit
Tricorn 10 Columns
Sinonimo/i:
Filtration assembly, Filtration kit
About This Item
Prodotti consigliati
Descrizione
Tricorn glass columns may be used with aqueous buffer solutions and detergents at pH conditions between 1 and 14, and with nearly all organic solutions commonly used in chromatography.
Caratteristiche
Ethylene propylene diene (EPDM) Polyethylene (PE)
Kit Contents: 5 x Top filter 5 x Bottom filter 5 x O-ring 7 x 1 mm (EPDM)
Tricorn 10 Columns
Wetted Material: Ethylene propylene diene (EPDM) Polyethylene (PE)
Produttore/marchio commerciale
Cytiva 29053612
Parametri
4 °C min. temp.
60 °C max. temp.
Compatibilità
Tricorn glass columns may be used with aqueous buffer solutions and detergents at pH conditions between 1 and 14, and with nearly all organic solutions commonly used in chromatography.
Descrizione generale
Tricorn columns incorporate modern design and high performance to increase the user friendliness and reliability of the chromatography column. Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures.
Caratteristiche e vantaggi
- High performance glass columns designed for reliable high resolution, reproducibility and durability
- New longer adaptor for increased adjustment of bed height
- Modern, integrated design – simple to assemble, pack, operate, and maintain
- Broad range of column dimensions – suitable for many different purification needs
- Safety first – glass tubes coated with plastic film to protect laboratory personnel if the column is dropped or operated outside its specifications
- Locking ring – prevents accidental compression of media bed
- Separate packing connector – allows simple column packing for a broad range of media
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
GE29053612 |