GBL702430
Grace Bio-Labs HybriSlip™ hybridization cover
L × W × thickness 24 mm × 30 mm × 0.25 mm, pkg of 100 ea
Sinonimo/i:
coverslide, hydrophobic coverslip, nuclease free coverslip, thin plastic coverslip
About This Item
Prodotti consigliati
Materiali
colorless
polycarbonate
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
Grace Bio-Labs 702430
Lungh. × largh. × spess.
24 mm × 30 mm × 0.25 mm
Descrizione generale
HybriSlips are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. HybriSlips remain flat and will not curl, even at high temperatures.
- RNase and DNase free
- Hydrophobic
- Disposable
- Will not chip or break
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
GBL702430-100EA | 4061832400969 |