GBL666384
Grace Bio-Labs Press-To-Seal silicone isolator, red silicone sheet
thickness 2.6 mm
Sinonimo/i:
Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier
About This Item
Prodotti consigliati
Materiali
red silicon
Sterilità
non-sterile
Caratteristiche
(Adhesive on both sides)
Confezionamento
pack of 5 ea
Produttore/marchio commerciale
Grace Bio-Labs 666384
Lungh. × largh.
180 mm × 130 mm
Spessore
2.6 mm
Categorie correlate
Descrizione generale
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
GBL666384-5EA | 4061832400846 |