GBL665384
Grace Bio-Labs silicone isolator sheet material
thickness 2.5 mm, (Adhesive on one side)
Sinonimo/i:
adhesive isolator, silicone blocker
About This Item
Codice UNSPSC:
41161502
NACRES:
NB.22
Prodotti consigliati
Materiali
red silicon
Sterilità
non-sterile
Caratteristiche
(Adhesive on one side)
Confezionamento
pack of 5 ea
Produttore/marchio commerciale
Grace Bio-Labs 665384
Lungh. × largh.
180 mm × 130 mm
Spessore
2.5 mm
Descrizione generale
Silicone isolator sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended.Silicone sheet material is also available in CultureWell “water-clear” silicone, which is highly recommended for sensitive cell culture applications.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
GBL665384-5EA | 4061832400532 |