Documenti fondamentali
EPM1282-9910
Platform for Eppendorf® New Brunswick Innova® 44/44R Incubator Shakers
for 2 L Erlenmeyer flasks, dedicated, non-Interchangeable, aluminum
About This Item
Codice UNSPSC:
41103816
NACRES:
NB.73
Materiali
aluminum
Caratteristiche
dedicated
for 2 L Erlenmeyer flasks
non-Interchangeable
Produttore/marchio commerciale
Eppendorf® M1282-9910 (Global)
Capacità
(6 x 2.8 Fernback flasks)
Categorie correlate
Descrizione generale
Platforms to fit the Eppendorf New Brunswick 44 and 44 R incubated shakers.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)