EPM1282-9904
Platform for Eppendorf® New Brunswick™ Innova® 44/44R Incubator Shakers
universal, non-Interchangeable, aluminum
About This Item
Codice UNSPSC:
41103816
NACRES:
NB.73
Prodotti consigliati
Materiali
aluminum
Caratteristiche
non-Interchangeable
universal
Produttore/marchio commerciale
Eppendorf® M1282-9904 (Global)
Misura
76 cm × 46 cm
Categorie correlate
Descrizione generale
Platforms to fit the Eppendorf New Brunswick 44 and 44 R incubated shakers.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc.
New Brunswick is a trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)