Documenti fondamentali
EPM1231-9933
Platform for Eppendorf® New Brunswick Innova® 3100 Water Bath Shaker
for 50 mL Erlenmeyer flasks, dedicated, interchangeable
About This Item
Codice UNSPSC:
41103816
NACRES:
NB.73
Materiali
stainless steel
Caratteristiche
dedicated
for 50 mL Erlenmeyer flasks
interchangeable
Produttore/marchio commerciale
Eppendorf® M1231-9933 (Global)
Lungh. × largh.
30.5 cm × 42 cm
Categorie correlate
Descrizione generale
Platform to fit the Eppendorf New Brunswick Innova 3100 water shaking waterbath.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)