EP5920755005
Adapter for Eppendorf® Rotor S-4xUniversal-Large
50mL conical tubes, 175-250 mL bottles and plates, 175-250 mL bottles and plates, rotor S-4xUniversal Large
Sinonimo/i:
Adapter for Eppendorf® S-4x Universal Large Rotor
About This Item
Codice UNSPSC:
41103911
NACRES:
NB.15
Caratteristiche
50mL conical tubes, 175-250 mL bottles and plates
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
5920755005
Compatibilità
rotor S-4xUniversal Large
Descrizione generale
Adapter to hold 50 mL conical tubes, 175-250 mL bottles and plates to be used with Eppendorf rotor S-4xUniversal. Supplied as a pack of 2 adapters.
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)