Documenti fondamentali
EP5804756002
Eppendorf® Adapter For 100 mL Rectangular Bucket
holds standard, Vacutainer, Falcon and micro test tubes 7-18 mL, pack of 2 ea
About This Item
Codice UNSPSC:
41103911
NACRES:
NB.82
Caratteristiche
holds standard, Vacutainer, Falcon and micro test tubes 7-18 mL
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Eppendorf® 5804756002
tecniche
centrifugation: suitable
Compatibilità
for use with 100 mL rectangular bucket
Descrizione generale
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Note legali
Eppendorf is a registered trademark of Eppendorf AG