Documenti fondamentali
EP5427735003
Eppendorf® Adapter
holds 1 x 9-15 mL, bore size (small), pack of 2 ea
About This Item
Codice UNSPSC:
41103911
NACRES:
NB.82
Caratteristiche
holds 1 x 9-15 mL
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Eppendorf® 5427735003
tecniche
centrifugation: suitable
Dimensioni foro
(small)
Compatibilità
for use with Rotor F-35-6-30 (small rotor bore)
Note legali
Eppendorf is a registered trademark of Eppendorf AG