Documenti fondamentali
EP0030128966
Eppendorf® Twin tec PCR plates, 384 wells
blue polypropylene plate, conical bottom colorless wells, pkg of 300 ea
About This Item
Codice UNSPSC:
41106300
NACRES:
NB.15
Materiali:
blue polypropylene plate
conical bottom colorless wells
Misura:
384 wells
Volume massimo del pozzetto:
45 μm
Volume d’esercizio:
-40 μL
Sterilità:
non-sterile
Tipo di binding:
low binding surface
Caratteristiche:
skirt
Materiali
blue polypropylene plate
conical bottom colorless wells
Sterilità
non-sterile
Caratteristiche
skirt
Confezionamento
pkg of 300 ea
Produttore/marchio commerciale
Eppendorf® 0030128966
tecniche
PCR: suitable
Misura
384 wells
Volume massimo del pozzetto
45 μm
Volume d’esercizio
-40 μL
Tipo di binding
low binding surface
Descrizione generale
“Eppendorf” Twin tec PCR plates with 384 wells are large scale PCR plates used in high throughput PCR workflows. They have rigid polycarbonate frames, polypropylene wells and raised well rims, ensuring a sturdy plate with effective sealing.
Applicazioni
Eppendorf “twin.tec” PCR Plates may be used in a wide variety of applications such as:
  • Standard PCR
  • Real time PCR
  • High throughput PCR
Caratteristiche e vantaggi
  • “Eppendorf” Twin tec PCR plates have polypropylene wells with a maximum volume of 40?L.
  • Enhanced rim design for efficient barcode scanning and automated workflows.
  • The plates can be stacked together enabling convenient storage and handling.
  • These Twin tec PCR plates are compatible with most 384- well thermocycler.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)